IPTG (isopropyl-β-D-thiogalactopyranoside) 99, ultrapure

Supplier: VWR Chemicals

Synonyms: IPTG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol, Isopropyl Beta-D-1-thiogalactopyranoside

J436-100GEA 234171 INR
J436-100G
IPTG (isopropyl-β-D-thiogalactopyranoside) 99, ultrapure
IPTG (Isopropyl-β-D-thiogalactopyranoside)
Formula: C₉H₁₈O₅S
MW: 238.31 g/mol
Boiling Pt: 402
Melting Pt: 109.5…110.5 °C
Storage Temperature: Freezer
MDL Number: MFCD00063273
CAS Number: 367-93-1
EINECS: 206-703-0

Order Now

Specification Test Results

Abs.@300 nm (5%, water) <0.13
Abs.@400 nm (5%, water) <0.06
Dioxane Test Pass
Melting Point 110 – 114 °C
Moisture (KF) <1%
pH (5%, water) @25 °C 5.0 – 7.0
Purity >99%
Solubility (5%, water) Pass
Specific Rotation (1%, water) -34.5 to -28.5°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR