CHEMSTAR™ Vacuum Pumps
Supplier: GARDNER DENVER
U1402N-50
ILMVU1402N-50EA 0 INR
ILMVU1402N-50
CHEMSTAR™ Vacuum Pumps
Pumps Vacuum Pumps
CHEMSTAR® pumps are built to withstand corrosive chemical vapor environments - durable performance over the long haul. CHEMSTAR® is designed to minimise the effects of harmful chemicals.
  • CHEMSTAR pump should not be operated continuously at pressures above 10 torr
  • Ultimate pressure measured with a trapped McCleod gauge
  • All single phase motors have overload protection
  • Conduit wiring installation required. No cord, plug or switch provided
Standard filter option possible if the exhaust flange is removed, but not chemically resistant.
Pump rpm: 525