Dry vacuum and chemical resistant diaphragm pumps (MPC)
Supplier: GARDNER DENVER
412522 412742-02 412543 412422-02 412443-02 412743 412722 412741 412711 412721 412781 412742 412782 412783 412943 412983 412722-30 412422-10 412443-10 412421-02 412021 412642 412744 414711
ILMV412522EA 0 INR
ILMV412522 ILMV412742-02 181-0515 ILMV412422-02 ILMV412443-02 ILMV412743 181-0335 ILMV412741 ILMV412711 ILMV412721 ILMV412781 ILMV412742 ILMV412782 ILMV412783 ILMV412943 ILMV412983 ILMV412722-30 ILMV412422-10 ILMV412443-10 ILMV412421-02 ILMV412021 ILMV412642 ILMV412744 ILMV414711
Dry vacuum and chemical resistant diaphragm pumps (MPC)
Pumps Vacuum Diaphragm Pumps
The MPC chemical duty diaphragm pumps use PTFE and other fluorinated plastics for the wetted parts to allow aggressive solvent and acid vapours to be pumped. If the pump should handle harsh chemical vapours, then the superior chemical resistant X2 version with higher grade seals and thicker coatings are the best choice. These are well known for quiet operation and running smoothness.
  • Different formats are available (1, 2, 4 head diaphragm pumps)
  • Low weight and compact design
  • Can be moved to different locations based on applications
They are availableMore Product Information