Dry vacuum and chemical resistant diaphragm pumps
Supplier: GARDNER DENVER
4000511-04
ILMV4000511-04EA 0 INR
ILMV4000511-04
Dry vacuum and chemical resistant diaphragm pumps
Pumps Vacuum Diaphragm Pumps
These are certified pumps for the evacuation of combustible atmospheres inside or outside of highly combustible environments. They are used for operating in potentially explosive atmospheres.
Certifications: Complies to 1999/92/EG (ATEX 137).