Dispersers, LABOR-PILOT 2000/4
Supplier: IKA JANKE KUNKEL
T055142 T054916 T055013 T061069 T054917
IKAAT055142EA 0 INR
IKAAT055142 IKAAT054916 IKAAT055013 IKAAT061069 IKAAT054917
Dispersers, LABOR-PILOT 2000/4
Homogenisers Homogenisers Bench
The LABOR-PILOT is a compact and multi-purpose inline laboratory dispersing machine. It is designed for solid-liquid mixing, homogenising, emulsifying, suspending and wet milling. The LABOR-PILOT 2000/4 is suitable for wide range of applications, particularly in the chemical, cosmetic, pharmaceutical, plastics, colours and food industries. It is uniquely designed with advanced technology and is available with six interchangeable modules.
  • Flexibility and ease of use: One machine suits for many applications and variety of processes
  • Together with inline
More Product Information