HPLC/UHPLC columns for small molecule analysis, HALO® C18, 2,7 µm
Supplier: HICHROM
92812-402 92812-602
HISM92812-402EA 0 INR
HISM92812-402 HISM92812-602
HPLC/UHPLC columns for small molecule analysis, HALO® C18, 2,7 µm
Chromatography Columns
HALO® HPLC and UHPLC columns are based on innovative fused-core particle technology. These particles are formed from a solid core and porous outer shell. HALO® 2,7 µm columns provide the efficiency and separation speed of sub 2 µm particles but at approximately half the back pressure for the same column length. This lower pressure means that HALO® columns can be used with conventional HPLC and LC-MS systems, as well as with UHPLC systems.
  • Ultra-fast separations
  • High resolution and high efficiency
  • Narrow particle size distribution
  • Lower backpressure than
More Product Information