Avantor® Hichrom, Connectors, Stainless Steel
Supplier: Avantor
HICHHI-752EA 0 INR
HICHHI-752
Avantor® Hichrom, Connectors, Stainless Steel
Fittings and Connectors
The female-female connector consists of stainless steel tubing (40 mm×1,588×0,254 mm (L×O.Ø×I.Ø)) with appropriate 1/16" nuts and ferrules and can be used to connect female endfitting guard and analytical columns and make similar connections.
  • ZDV unions are rated to 20000 psi
The ZDV male-male connectors (unions) have internal threads and provide zero dead volume connections.