Avantor® Hichrom PEEKsil™, Fused Silica Tubing
Supplier: Avantor
HICHHI-1097EA 0 INR
HICHHI-1097
Avantor® Hichrom PEEKsil™, Fused Silica Tubing
Tubing Chromatography Tubing
The maximum operating temperature is 100 °C.
  • pH range 0 to 10
  • Used for micro and nano scale HPLC and capillary electrophorosis
Manufactured from synthetic fused silica with a polyimide coating.