Avantor® Hichrom HI-1 MS, GC Columns

Supplier: Avantor
HI16-25-025-15G5EA 51610 INR
HI16-25-025-15G5 HI16-25-025-30G5 HI16-25-025-60G5 HI16-53-100-30G5 HI16-18-018-10 HI16-18-018-20 HI16-18-018-40 HI16-18-018-50 HI16-18-018-60 HI16-18-020-10 HI16-18-020-20 HI16-18-020-40 HI16-18-020-50 HI16-18-020-60 HI16-18-036-20 HI16-18-036-40 HI16-18-036-60 HI16-18-040-10 HI16-18-040-20 HI16-18-040-40 HI16-18-040-50 HI16-18-040-60 HI16-20-010-10 HI16-20-010-20 HI16-20-010-40 HI16-20-010-50 HI16-20-010-60 HI16-20-020-10 HI16-20-020-20 HI16-20-020-40 HI16-20-020-50 HI16-20-020-60 HI16-20-033-12 HI16-20-033-25 HI16-20-033-30 HI16-20-033-50 HI16-20-033-60 HI16-20-040-10 HI16-20-040-20 HI16-20-040-40 HI16-20-040-50 HI16-20-040-60 HI16-20-100-10 HI16-20-100-20 HI16-20-100-40 HI16-20-100-50 HI16-20-100-60 HI16-25-010-15 HI16-25-010-30 HI16-25-010-50 HI16-25-010-60 HI16-25-025-10 HI16-25-025-105 HI16-25-025-15 HI16-25-025-30 HI16-25-025-50 HI16-25-025-60 HI16-25-050-10 HI16-25-050-105 HI16-25-050-15 HI16-25-050-30 HI16-25-050-50 HI16-25-050-60 HI16-25-100-10 HI16-25-100-105 HI16-25-100-15 HI16-25-100-30 HI16-25-100-50 HI16-25-100-60 HI16-32-010-10 HI16-32-010-105 HI16-32-010-15 HI16-32-010-30 HI16-32-010-50 HI16-32-010-60 HI16-32-025-10 HI16-32-025-105 HI16-32-025-15 HI16-32-025-30 HI16-32-025-50 HI16-32-025-60 HI16-32-050-10 HI16-32-050-105 HI16-32-050-15 HI16-32-050-30 HI16-32-050-50 HI16-32-050-60 HI16-32-100-10 HI16-32-100-105 HI16-32-100-15 HI16-32-100-30 HI16-32-100-50 HI16-32-100-60 HI16-32-150-10 HI16-32-150-105 HI16-32-150-15 HI16-32-150-30 HI16-32-150-50 HI16-32-150-60 HI16-53-010-10 HI16-53-010-15 HI16-53-010-30 HI16-53-025-15 HI16-53-025-30 HI16-53-025-50 HI16-53-025-60 HI16-53-050-10 HI16-53-050-15 HI16-53-050-30 HI16-53-050-50 HI16-53-050-60 HI16-53-100-10 HI16-53-100-15 HI16-53-100-30 HI16-53-100-50 HI16-53-100-60 HI16-53-100-75 HI16-53-150-10 HI16-53-150-15 HI16-53-150-30 HI16-53-150-50 HI16-53-150-60 HI16-53-150-75 HI16-05-005-2 HI16-05-005-2.5 HI16-05-005-5 HI16-10-010-10 HI16-10-010-15 HI16-10-010-5 HI16-10-020-10 HI16-10-020-15 HI16-10-020-5 HI16-10-040-10 HI16-10-040-15 HI16-10-040-5 HI16-15-015-10 HI16-15-015-15 HI16-15-015-20 HI16-15-015-30 HI16-15-015-5
Avantor® Hichrom HI-1 MS, GC Columns
Chromatography Columns
Really versatile general purpose apolar phase for a wide range of MS applications, including (for example): PCBs, simulated distillation, gases, essential oils, hydrocarbons analysis, pesticides, semivolatiles, oxygenates. HI-1 MS is an MS grade low-bleeding crossbonded column and assure a long lifetime even under high temperature limits.

  • Phase composition: Crossbonded 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - MS grade
  • Long lifetime
  • Equivalent to USP phases G1, G2, G38

Max temperature limits may change depending on the film thickness.
Order Now

SPECIFICATIONS

Environmentally Preferable
Phase name HI-1 MS
Phase composition 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
Polarity Apolar stationary phase
Crossbond Yes
Temperature limit 350 °C
USP code G1, G2, G9, G38

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR