Ultra-low temperature freezers, –86 to –40 ℃
Supplier: HAIER MEDICAL LABORATORY PRO
DW-86W420 DW-86L959 DW-86L338 DW-86L486 DW-86L628 DW-86L728 DW-86L828 DW-86W100 DW-86L729 DW-86L388A DW-86L490J DW-86L728J DW-86L490J/SS DW-86L628/SS
HAIEDW-86W420EA 0 INR
HAIEDW-86W420 HAIEDW-86L959 HAIEDW-86L338 HAIEDW-86L486 HAIEDW-86L628 HAIEDW-86L728 HAIEDW-86L828 HAIEDW-86W100 HAIEDW-86L729 HAIEDW-86L388A HAIEDW-86L490J HAIEDW-86L728J HAIEDW-86L490J/SS HAIEDW-86L628/SS
Ultra-low temperature freezers, –86 to –40 ℃
Freezers Ultra Low Temperature Freezers
Upright ultra low temperature freezers, for long term storage of biological products, including viruses, germs, erythrocytes, leucocytes and cutis. Units have a lockable handle with a key that is unique to that unit, preventing any access to precious samples, there is also a space for a padlock for extra security.
  • Reliable performance: Cabinet temperature can reach –86 ℃ with temperature uniformity within 6 ℃ (or 5 ℃ for model WD-86L728J)
  • Energy saving: Four independent inner doors and four layer sealing structure ensures good heat insulation to reduce
More Product Information