Forceps
Supplier: HACH
2141100
HACH2141100EA 0 INR
HACH2141100
Forceps
Forceps
Stainless steel.
  • Tips are rounded to prevent damage
  • Easy to use because of slip-proof plastic finger tips
  • Autoclavable