Temperature logger, Squirrel OQ610 Series
Supplier: GRANT INSTRUMENTS
OQ610-S
GRANOQ610-SEA 0 INR
GRANOQ610-S
Temperature logger, Squirrel OQ610 Series
Data Loggers
This logger is suitable for a wide range of temperature recording applications in industry, research and development.
  • Six channels for use with a wide range of K or T type thermocouple probes
  • Simple three button operation via built-in display or remote control from a PC
  • Automatic start and stop at specific times or temperature levels
  • Fast sample rates for fast process times: Up to eight samples per second
  • Automatic cure calculation in process applications
  • Slowest process time: once every two hours per channel
  • Eight number of runs can be stored