Avantor® ACE® Generix® C18 & C18(2), Analytical HPLC Columns
Supplier: Avantor
GEN-5C18-1546EA 42190 INR
GEN-5C18-1546 GEN-5C18-2546 ACETGEN-3C18-0502 ACETGEN-3C18-1002 ACETGEN-3C18-1046 ACETGEN-3C18-1502 ACETGEN-3C18-1546 ACETGEN-5C18-1046 GEN-5C18-GD GEN-3C182-1046 GEN-3C182-1546 GEN5C1821546/3 GEN5C1822546/3 GEN-5C182-2546 GEN-5C182-1546 GEN3C1820502/3 GEN3C1821046/3 GEN3C1821546/3 GEN-C182-GD2 GEN-C182-GD GEN-3C182-0502 GEN-3C182-7502 GEN-3C182-1002 GEN-3C182-1202 GEN-3C182-1502 GEN-3C182-0503 GEN-3C182-7503 GEN-3C182-1003 GEN-3C182-1203 GEN-3C182-1503 GEN-3C182-0504 GEN-3C182-7504 GEN-3C182-1004 GEN-3C182-1204 GEN-3C182-1504 GEN-3C182-0546 GEN-3C182-7546 GEN-3C182-1246 GEN-5C182-0502 GEN-5C182-7502 GEN-5C182-1002 GEN-5C182-1202 GEN-5C182-1502 GEN-5C182-2502 GEN-5C182-0503 GEN-5C182-7503 GEN-5C182-1003 GEN-5C182-1203 GEN-5C182-1503 GEN-5C182-2503 GEN-5C182-0504 GEN-5C182-7504 GEN-5C182-1004 GEN-5C182-1204 GEN-5C182-1504 GEN-5C182-2504 GEN-5C182-0546 GEN-5C182-7546 GEN-5C182-1046 GEN-5C182-1246 GEN-3C18-GD
Avantor® ACE® Generix® C18 & C18(2), Analytical HPLC Columns
Chromatography Columns
These high performance HPLC columns contain high purity spherical silica.
  • Ultra-narrow particle size distribution for excellent column efficiency
  • Sharp, highly symmetrical peaks for acidic, basic and neutral analytes
  • Excellent column to column and batch to batch reproducibility
Each column is individually tested and supplied with a test chromatogram.
Two generations of Avantor® ACE® Generix® C18 exist: C18 and C18(2) they differ in pore size, carbon load and surface area.