Pipette Controllers
Supplier: ACCUMAX
PH01-R PH01-B PH01-O PH01-G
FINEPH01-REA 9839 INR
FINEPH01-R FINEPH01-B FINEPH01-O FINEPH01-G
Pipette Controllers
Pipettes Pipette Controllers
These pipette controllers features lightweight construction for constant ease of use over prolonged usage.
  • Powerful pump - 25 ml pipette in 4 seconds
  • Eco-friendly rechargeable NiMH batteries, offering upto 8 hours of continuous output, 2 to 3 hours to charge fully
  • Easily replaceable batteries
  • Autoclavable tip cone for avoiding cross-contamination
  • Available in vibrant colors
Unique nose cone angle for enhanced flexibility in operation, even when in fume hood. Intelligent charger prevents overcharging/heating of batteries and can be used separately, andMore Product Information