Avantor® ACE® Excel® C18, HPLC/UHPLC Columns, Analytical, 5 µm
Supplier: Avantor
EXL-121-1002UEA 40960 INR
EXL-121-1002U EXL-121-1003U EXL-121-1046U EXL-121-1203U EXL-121-1246U EXL-121-1502U EXL-121-1503U EXL-121-1546U EXL-121-2502U EXL-121-2503U EXL-121-2546U EXL-121-0502U EXL-121-0503U EXL-121-0546U EXL-121-7502U EXL-121-7503U EXL-121-7546U
Avantor® ACE® Excel® C18, HPLC/UHPLC Columns, Analytical, 5 µm
Chromatography Columns
Avantor® ACE® C18 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (1.7 to 5 µm) and complementary phases
  • Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range ofMore Product Information