Floor marking and striping rolls, DuraStripe® Supreme V
Supplier: ERGOMAT
DSV2100Y
ERGODSV2100YEA 0 INR
ERGODSV2100Y
Floor marking and striping rolls, DuraStripe® Supreme V
Signs
DuraStripe® is the easiest, fastest and least expensive floor marking system for industrial floors. The durability and vast size of the DuraStripe program makes it the only total floor marking solution available on the market today. Economic than painting the floors.
  • Ideal for less than perfect floors
  • Simple and easy peel-&-stick installation
  • No dry time, cure time, fumes or messy clean up to limit productivity
The Supreme V combines the adhesive from the original DuraStripe which is the superior choice under non ideal floor conditions along withMore Product Information