1-Naphtholbenzeine, OR, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Warning

Synonyms: alpha-Naphtholbenzein, p-Naphtholbenzein, α-Naphtholbenzein, p-Naphtholbenzeine, 4-(α-(4-Hydroxy-1-naphthyl)benzylidene)naphthalen-1(4H)-one, 1-Naphtholbenzein, Bis(4-hydroxy-1-naphthyl)phenylmethanol, 4-Hydroxy-α-(4-hydroxynaphthyl)-α-phenylnaphthalene-1-methanol, α-Naphtholbenzeine

E121-54EA 0 INR
E121-54
1-Naphtholbenzeine, OR, Macron Fine Chemicals™
1-Naphtholbenzeine
Formula: C₂₇H₁₈O₂
MW: 374.44 g/mol
Melting Pt: 243…245 °C
Storage Temperature: Ambient
MDL Number: MFCD00078492
CAS Number: 145-50-6
EINECS: 205-656-3

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Solubility Passes Test
Identification (by IR) Passes Test
Visual Transition Intervals - pH...(Orange) 8.2 Passes Test
Visual Transition Intervals - pH...(Blue) 10.0 Passes Test
Appearance (brownish-red powder)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR