External Dimensions: 565 x 510 x 435 mm Internal Dimensions: 444 x 388 x 302 mm Dry-Ice Mass: 33.1 kg Payload Volume: 16.8 L Duration: 96+ 1 * 1 items
Supplier: DGP INTELSIUS
External Dimensions: 565 x 510 x 435 mm Internal Dimensions: 444 x 388 x 302 mm Dry-Ice Mass: 33.1 kg Payload Volume: 16.8 L Duration: 96+ 1 * 1 items
Learn more About VWR
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com