Intelsius® BioTherm™ Dry Ice Shippers
Supplier: DGP INTELSIUS
BT074 BT041 BT049 BT020 BT050
DGINBT074EA 0 INR
DGINBT074 DGINBT041 DGINBT049 DGINBT020 DGINBT050
Intelsius® BioTherm™ Dry Ice Shippers
Mailers and Shipping Containers
Intelsius® Biotherm packaging systems are regulatory compliant to IATA, ADR, and 49 CFR (DOT) transport regulations. These shippers utilise dry ice to maintain sample integrity during shipments from 24 to 240 hours.
  • Regulatory compliant
  • Dry ice shippers
  • Compliant markings on boxes
  • Variety of internal volumes
Intelsius® Biotherm shipping systems are pre-qualified to maintain sample integrity during shipments from 24 to 240 hours against demanding high-performance dry ice temperature (2 to 8 °C profiles and 15 to 25 °C profiles). The range is tested andMore Product Information