Safety spectacles, Uvex Ultra-spec®
Supplier: HONEYWELL SAFETY PRODUCTS
S0300
DALLS0300EA 0 INR
DALLS0300
Safety spectacles, Uvex Ultra-spec®
Safety Glasses
Economical safety glasses feature a 100% polycarbonate wrap around unilens design.
  • Clear lenses and frames
  • Adjustable temple length
  • Side shields add extra protection without compromising visibility
  • Ultra-dura® and Uvextreme® lens coating provides superior scratch and chemical resistance
Weight: 44 g
Order Now
SPECIFICATIONS
Environmentally Preferable