E.Z.N.A.® Endo-Free plasmid kits

Supplier: OMEGA BIO-TEK

D6926-03 D6948-01 D6926-01 D6926-04 D6915-03 D6948-00 D6915-00 D6926-00
D6926-03EA 0 INR
D6926-03 D6948-01 D6926-01 D6926-04 D6915-03 D6948-00 D6915-00 D6926-00
E.Z.N.A.® Endo-Free plasmid kits
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

Plasmid isolated with traditional purification procedures normally contain high levels of endotoxins that can significantly interfere with transfection experiments downstream. The E.Z.N.A.® Endo-Free Plasmid kits integrate an efficient endotoxin removal step into the plasmid purification procedure to produce high quality transfection grade plasmid.

Order Now

SPECIFICATIONS

Description E.Z.N.A.® Fastfilter® Endo-Free Plasmid Maxi kit E.Z.N.A.® Endo-Free Plasmid Mini Kit I E.Z.N.A.® Endo-Free Plasmid Midi Kit E.Z.N.A.® Fastfilter® Endo-Free Plasmid Maxi Kit
Environmentally Preferable
Application
Target molecule Plasmid Plasmid Plasmid Plasmid
Refine target molecule Plasmid (endotoxin-free) Plasmid (endotoxin-free) Plasmid (endotoxin-free) Plasmid (endotoxin-free)
Sample type Bacteria Bacteria Bacteria Bacteria
Method / Format Column (silica-based) Column (silica-based) Column (silica-based) Column (silica-based)
Prep size Maxi Mini Midi Maxi
Sample size 50 - 200 ml 1 - 5 ml 15 - 50 ml 50 - 200 ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR