Simulated gastric fluid (without enzyme) concentrate for dilution to (NaCl 2 g/L), DILUT-IT® Dissolution Media Concentrate, J.T.Baker®

Supplier: Avantor
Danger

D020-02EA 0 INR
D020-02
Simulated gastric fluid (without enzyme) concentrate for dilution to (NaCl 2 g/L), DILUT-IT® Dissolution Media Concentrate, J.T.Baker®
Simulated gastric fluid (without enzyme)
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use 1.9 - 2.1 g/l
Assay, Sodium Chloride (NaC₁) (at 26x dilution) 1.9 - 2.1 g/l
pH at 25°C at 26x Dilution 1.15 - 1.25
Does not contain enzyme


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR