Compression screw fittings
Supplier: COWIE
PC701.12 PC701.075.16 PC701.075.19 PC701.08 PC701.100.08 PC701.100.12 PC701.100.14 PC701.100.16 PC701.075.12 PC701.100.25 PC701.075.08 PC701.14 PC701.16 PC701.19 PC701.25 PC701.100.19 PC701.025 PC701.003 PC701.006 PC701.008 PC701.012 PC701.014 PC701.075.14 PC701.019 PC701.025.03 PC701.025.06 PC701.03 PC701.050.06 PC701.050.08 PC701.050.12 PC701.050.14 PC701.06 PC701.016 PC703.06 PC703.08 PC703.12 PC701.250 PC701.750 PC701.625 PC701.560 PC701.500 PC701.1000 PC701.312 PC701.0560 PC701.100.750 PC701.100.625 PC701.100.560 PC701.100.500 PC701.100.1000 PC701.0750 PC701.075.750 PC701.075.625 PC701.075.560 PC701.075.500 PC701.0625 PC701.0500 PC701.050.560 PC701.050.500 PC701.050.312 PC701.050.250 PC701.0312 PC701.0250 PC701.025.250 PC701.025.125 PC701.0125 PC701.01000 PC701.075.312
COWIPC701.12EA 0 INR
COWIPC701.12 COWIPC701.075.16 COWIPC701.075.19 COWIPC701.08 COWIPC701.100.08 COWIPC701.100.12 COWIPC701.100.14 COWIPC701.100.16 COWIPC701.075.12 COWIPC701.100.25 COWIPC701.075.08 COWIPC701.14 COWIPC701.16 COWIPC701.19 COWIPC701.25 COWIPC701.100.19 COWIPC701.025 COWIPC701.003 COWIPC701.006 COWIPC701.008 COWIPC701.012 COWIPC701.014 COWIPC701.075.14 COWIPC701.019 COWIPC701.025.03 COWIPC701.025.06 COWIPC701.03 COWIPC701.050.06 COWIPC701.050.08 COWIPC701.050.12 COWIPC701.050.14 COWIPC701.06 COWIPC701.016 COWIPC703.06 COWIPC703.08 COWIPC703.12 COWIPC701.250 COWIPC701.750 COWIPC701.625 COWIPC701.560 COWIPC701.500 COWIPC701.1000 COWIPC701.312 COWIPC701.0560 COWIPC701.100.750 COWIPC701.100.625 COWIPC701.100.560 COWIPC701.100.500 COWIPC701.100.1000 COWIPC701.0750 COWIPC701.075.750 COWIPC701.075.625 COWIPC701.075.560 COWIPC701.075.500 COWIPC701.0625 COWIPC701.0500 COWIPC701.050.560 COWIPC701.050.500 COWIPC701.050.312 COWIPC701.050.250 COWIPC701.0312 COWIPC701.0250 COWIPC701.025.250 COWIPC701.025.125 COWIPC701.0125 COWIPC701.01000 COWIPC701.075.312
Compression screw fittings
Fittings and Connectors
Compression fittings are manufactured from pure PTFE and include a PEEK olive to give a secure grip.
  • Excellent chemical resistance
  • Excellent temperature resistance
Compression fittings are used with PTFE reactor lids to hold items such as temperature probes and sampling tubes securely in place.
PTFE white plugs are used to seal a compression fitting without the need to remove the compression fitting from the reactor lid. They are chemically inert, reduce contamination and simplify cleaning and are non-scratchable.