Beakers with handle
Supplier: BURKLE
7110-2000 7110-1000
BURK7110-2000EA 0 INR
BURK7110-2000 BURK7110-1000
Beakers with handle
Beakers
PP, translucent.
  • Shatterproof
  • Autoclavable
  • Good chemical resistance
With open handle. Beakers can be stacked and therefore save space during storage.