Rotary evaporators, Rotavapor® R-300
Supplier: BUCHI
11SR300152VP21
BUCH11SR300152VP21EA 0 INR
BUCH11SR300152VP21
Rotary evaporators, Rotavapor® R-300
Evaporators Rotary Evaporators
Its modular design allows the extension of the Rotavapor® R-300 to a fully integrated system where a central interface regulates each component.
  • Comfortable flask and vapor duct removal thanks to easy to use Combi-Clip
  • User-friendly operation due to ergonomic electronic or manual lift
  • Immediate adjustment of heating temperature and rotation speed with intuitively positioned control knobs
  • Easy plug & play extension to a fully integrated system including a central interface, vacuum pump and recirculating chiller
  • Precise operation control with integrated
More Product Information