Avantor® ACE® UltraCore BIO C4, Analytical HPLC/UHPLC Columns
Supplier: Avantor
BIO-251-521GEA 59040 INR
BIO-251-521G BIO-251-0521 BIO-251-1021 BIO-251-1521 BIO-251-530G BIO-251-0530 BIO-251-1030 BIO-251-1530 BIO-251-546G BIO-251-0546 BIO-251-1046 BIO-251-1546 BIO-351-521G BIO-351-0521 BIO-351-1021 BIO-351-1521 BIO-351-530G BIO-351-0530 BIO-351-1030 BIO-351-1530 BIO-351-546G BIO-351-0546 BIO-351-1046 BIO-351-1546
Avantor® ACE® UltraCore BIO C4, Analytical HPLC/UHPLC Columns
Chromatography Columns
Avantor® ACE® UltraCore BIO C4 are high performing solid-core (core-shell) particle columns, designed for a range of biomolecule applications. With both 300 Å and 500 Å pore sizes available, optimum pore size can be chosen. This phase is well suited to the analysis of hydrophobic proteins over 5 kDa molecular weight.
  • Wide pore, solid-core combination for excellent peak shape & resolution of large molecules
  • High purity, base deactivated silica with excellent reproducibility
  • Butyl ligand well established in RP protein applications
These stainlessMore Product Information