BeneSphera™, Syringes
Supplier: BENESPHERA
16007470 16007472 16007473 16007474 16007475 16007476 16007477
BENP16007470EA 0 INR
BENP16007470 BENP16007472 BENP16007473 BENP16007474 BENP16007475 BENP16007476 BENP16007477
BeneSphera™, Syringes
Syringes Syringes, Medical
The needle point are CNC machine grinded to ensure uniform sharpness and minimal discomfort. This is achieved through high precision grinding, ultrasonic cleaning and siliconisation of surgical grade SS 304 canulae.
Disposable hypodermic syringes and needles manufactured from strictly selected high quality raw material, processed on fully automatic machines.
Certifications: Syringes specificatlons are as per ISO 7888, IS-10250, IS-10258. Colours are as per ISO 6009 for gauge specification. Needles specification are as per ISO 7864 and IS-10654.
Order Now
SPECIFICATIONS
Environmentally Preferable