BeneSphera™ Electrolyte Analysers - (Na/K/iCa/pH/HCO3/Cl)
Supplier: BENESPHERA
16007386 16007448 16007449 16007450 16007451 16007452 16007453 16007455 16007459 16007462 16007463 16007464 20001983 20001988 20002006 20002024
BENP16007386EA 8343 INR
BENP16007386 BENP16007448 BENP16007449 BENP16007450 BENP16007451 BENP16007452 BENP16007453 BENP16007455 BENP16007459 BENP16007462 BENP16007463 BENP16007464 BENP20001983 BENP20001988 BENP20002006 BENP20002024
BeneSphera™ Electrolyte Analysers - (Na/K/iCa/pH/HCO3/Cl)
Clinical Diagnostic Supplies Clinical Analysers
Electrolyte analyser allows results of 6 different parameters Na, K, iCa, PH, Li and Cl with a single sample.
  • 70 different in-built combinations from choice of six parameters (No, K, iCa, Li, a, pH) which can be upgraded on spot with universal reagent pack, QC and daily cleaner
  • Intelligent reagent pack with electronic chip ensures no generic possibility. The analyser is a completely closed system
  • Electrodes are entirely maintenance free. There is no requirement to refill IFS till the life span of the electrode
  • Maximum 700000+ sample storage capacity
More Product Information