BeneSphera™ T4 MW ELISA - Novo

Supplier: BENESPHERA

15001539
BENP15001539EA 3274.2 INR
BENP15001539
BeneSphera™ T4 MW ELISA - Novo
Assays ELISAs

BeneSphera™ T4 Novo Elisa kit is for quantitative determination of T4 (thyroxine) hormone from human serum and plasma.


  • Assay time: 1 hour 30 min
  • Expected range: male: 4,4 - 10,8 µg/dl, female: 4,8 - 11,6 µg/dl
  • Sensitivity: 0,4 µg/dl
  • Specificity: No interference found, a total of 131 samples found to have high degree of correlation with T4 calibrators provided


Enzyme Immunoassay for the quantitative determination of T4 Hormone concentration in human serum/plasma - for invitro diagnostics use only.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR