BeneSphera™ HIV 1/2 Ab 3.0 Ultra

Supplier: BENESPHERA

15001535
BENP15001535EA 1436.85 INR
BENP15001535
BeneSphera™ HIV 1/2 Ab 3.0 Ultra
Assays Immunological Assays

Rapid card tests (lateral flow) for detection of HIV antibodies [HIV 1 (including subgroup O) and HIV 2].


  • Detects and differentiates HIV 1 & 2
  • Detects total antibodies (IgG, IgM and IgA)
  • Detects antibodies to HIV 1 - O Subgroup
  • Sample Type: Serum/Plasma/Whole Blood
  • Finger prick can be used
  • Sensitivity: 100%
  • Specificity: 99,92%


BeneSphera™ HIV 1/2 Antibody 3.0 Ultra Rapid Card test is a rapid and visual immunochromatographic assay, intended for the qualitative detection of antibodies against HIV 1 (including group O) and/or HIV 2 from human whole blood, serum or plasma.


Certifications: National Institute of Biologicals (MoHFW, Govt. of India).

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR