BeneSphera™, HIV½ Ab 3.0 Rapid Test
Supplier: BENESPHERA
15001268 15001269 15001316
BENP15001268EA 0 INR
BENP15001268 BENP15001269 BENP15001316
BeneSphera™, HIV½ Ab 3.0 Rapid Test
Clinical Diagnostic Supplies Clinical Reagents and Kits
BeneSphera™ HIV 1/2 Ab 3.0 Rapid Test is a third generation test for differential detection of HIV-1 and HIV-2 antibodies ( IgG, IgM and IgA).
  • Target Marker: Gp41, Gp120, P24 for HIV 1 and Gp36 for HIV 2
  • Subtype O and C detectable
Order Now
SPECIFICATIONS
Environmentally Preferable