BeneSphera™ H-33S Lyse, 3 Part Differential
Supplier: BENESPHERA
15001300
BENP15001300EA 4635 INR
BENP15001300
BeneSphera™ H-33S Lyse, 3 Part Differential
Clinical Diagnostic Supplies Clinical Reagents and Kits
For determination of HB, counting and sizing of lucocytes in hematology analyser.
  • Best fit for 3 part hematology analyser
  • Storage at RT
  • Long shelf life
Caution: Avoid the contact with eyes and skin.