Benesphera™ EZ cleaner, 3 part differential analyser
Supplier: BENESPHERA
15001083
BENP15001083EA 1112.4 INR
BENP15001083
Benesphera™ EZ cleaner, 3 part differential analyser
Clinical Diagnostic Supplies Clinical Reagents and Kits
For Bath before shut down in hematology analyzer
  • Best Fit for 3 Part Hematology analyzer
  • Storage at RT
  • Long Shelf Life
Avoid the contact with eyes & Skin