BeneSphera™ Rinse, 3 Part Differential
Supplier: BENESPHERA
15001082
BENP15001082EA 1652 INR
BENP15001082
BeneSphera™ Rinse, 3 Part Differential
Clinical Diagnostic Supplies Clinical Reagents and Kits
Hematology rinse solution PH 7,00 for 3 part hematology analyser.
  • Best fit for 3 part hematology analyser
  • Storage at RT
  • Long shelf life
Caution: Avoid the contact with eyes and skin.