VWR® Culture Media, Ready to Use, in Bottles
Supplier: VWR Chemicals
AX021131EA 0 INR
AX021131 AX021135 AX021116 AX021109 AX021320 AX021162 AX022229 AX022279 AX021117 AX022258 AX022205 AX029178 AX029148 AX021102 AX021103 AX021133 AX021112 AX021126 AX021146 AX021190 AX021230 AX021255 AX021106 AX021118 AX021128 AX021130 AX021154 AX021169 AX021178 AX021182 AX021187 AX021191 AX021197 AX021203 AX021205 AX021206 AX021233 AX021235 AX021243 AX021250 AX021257 AX021258 AX021259 AX021261 AX021263 AX021264 AX021283 AX021291 AX021301 AX021305 AX021315 AX021317 AX021323 AX021324 AX021330 AX022206 AX022219 AX022222 AX022232 AX022241 AX022245 AX022255 AX022281 AX022284 AX022292 AX022293 AX029102 AX029124 AX029128 AX029129 AX029167 AX029174 AX029176 AX029179 AX029189 AX021111 AX021346 AX021361 AX021366 AX021367 AX021370 AX021373 AX021374 AX021378 AX021372 AX021380 AX021383 AX021396 AX021398 AX021399 AX021402 AX021404 AX021405 AX021406 AX021408 AX021409 AX021411 AX021412 AX029198 400442ZA 400304ZA 401312ZA 301114ZA 880304ZA 880442TA 880442ZA 881122TA 881122ZA AX021415 301112TA 471310TH 881120TA 300884TA 572512ZA AX021198 AX011201 011312TA 555570ZA
VWR® Culture Media, Ready to Use, in Bottles
Media for Microbiology Prepared Media for Microbiology
Ready to use culture media for microbiological test requirements in the food industry, water, environmental control and in the pharmaceutical industry. The range is manufactured from high quality raw materials. All the culture media are manufactured using selected dehydrated bases, and comply with various standards. All media are produced using the latest technology and are subject to strict quality controls carried out by qualified microbiologists.
  • Storage at room temperature for selected media
  • Supplied in glass bottles with different screw caps
  • Certificate of Analysis guarantees product conformity
*Chromocult® is a registered trademark of Merck KGaA, Darmstadt, Germany. This ready to use medium is made using only Chromocult® dehydrated media produced by Merck KGaA and Agar.