Disinfectant for incubators, Incuwater-Clean™
Supplier: PanReac AppliChem
A5219.0100
A5219.0100EA 0 INR
A5219.0100
Disinfectant for incubators, Incuwater-Clean™
Cleaning Agents Disinfectants
The water required to create the humidity is a potential source of contamination which can disperse into the incubator. Incuwater-Clean™ is an effective, non toxic, non volatile disinfectant that does not damage stainless steel or copper trays.
  • For the prevention of microbial growth in incubator water baths
  • In concentrate format (100X) – use 50 ml per 5 litres of water
  • Economical – replace bath contents with sterile water and dose with Incuwater-Clean™ every 2 to 4 weeks
Caution: Use biocides safely. Always read the label and product information before use.