Trypsin

Supplier: PanReac AppliChem

A3964.0005
A3964.0005EA 0 INR
A3964.0005
Trypsin
Enzymes

Order Now

SPECIFICATIONS

Enzyme name Trypsin
Environmentally Preferable
Source
Species
MDL No. MFCD00082094
CAS No. 9002-07-7
Storage conditions Refrigerator
Enzyme activity 2.500 U/mg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR