Ammonium sulphate

Supplier: PanReac AppliChem

Synonyms: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

A3485.5000
A3485.5000EA 0 INR
A3485.5000
Ammonium sulphate
Ammonium sulphate
Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Melting Pt: 336…339 °C
Density: 1.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR