VWR®, Smoke Point standard
Supplier: VWR Chemicals
99125.230EA 0 INR
99125.230
VWR®, Smoke Point standard
Standards Petroleum Industry Standards
Fuel testing Certified Reference Material for the calibration and verification of laboratory instruments used in testing petroleum and derivative products for Smoke Point ASTM D1322 / IP 57.
  • Certified in accordance with ASTM / EN or IP test method protocols
  • Fully traceable to international standards
  • Tamper-evident security packaging
Certifications: Fully certified in accordance with ISO 17034 under accreditation by UKAS.