Water, BAKER ANALYZED® LC-MS, J.T.Baker®

Supplier: Avantor

Synonyms: Hydrogen oxide

M.O.C. Category NI

9831-03EA 15361.62 INR
9831-03 9831-02 9831-P01 9831-P2.5 9831-P04 9831-88
Water, BAKER ANALYZED® LC-MS, J.T.Baker®
Water

For use in liquid chromatography and mass spectrometry.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use Passes Test
Appearance Passes Test
Gradient Elution Test (a.u.) - 254 nm ≤ 0.002
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤ 0.1 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤ 0.2 ppb
Color (APHA) ≤ 10
Residue after Evaporation ≤ 1.0 ppm
LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine) ≤ 50 ppb
Trace Impurities - Aluminum (Al) ≤ 50.0 ppb
Trace Impurities - * Calcium (Ca) ≤ 50.0 ppb
Trace Impurities - Iron (Fe) ≤ 50.0 ppb
Trace Impurities - Lithium (Li) ≤ 30.0 ppb
Trace Impurities - Magnesium (Mg) ≤ 50.0 ppb
Trace Impurities - Nickel (Ni) ≤ 30.0 ppb
Trace Impurities - Potassium (K) ≤ 50 ppb
Trace Impurities - * Sodium (Na) ≤ 50.0 ppb
* May change over time due to extraction from glass container.
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR