Sulphuric acid 95 - 98% 95,0-98,0 NF, multi-compendial, J.T.Baker®

Supplier: Avantor
Danger


M.O.C. Category R

9671-02EA 9507.28 INR
9671-02 9671-03 9671-26
Sulphuric acid 95 - 98% 95,0-98,0 NF, multi-compendial, J.T.Baker®
Sulphuric acid

GMP Manufactured Product Meets B.P. Chemical Specifications Meets E.P. Chemical Specifications Meets N.F. Requirements

Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: ∼335 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II
Merck Index: 14,08974

Order Now

Specification Test Results

GMP Manufactured Product
Meets N.F. Requirements
Meets E.P. Chemical Specifications
Meets B.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical Passes Test
NF - Identification Passes Test
NF - Residue on Ignition ≤ 0.005 %
NF - Chloride (Cl) ≤ 0.005 %
NF - Arsenic (As) ≤ 1 ppm
NF - Reducing Substances Passes Test
NF - Assay (as H₂SO₄) (by acidimetry) 95.0 - 98.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Chloride (Cl) ≤ 50 ppm
EP/BP - Nitrate (NO₃) Passes Test
EP/BP - Iron (Fe) ≤ 25 ppm
EP/BP - Assay (H₂SO₄) 95.0 - 100.5 %
No Class 1,2,3 or other solvents are used or produced in the = 5 ppm
manufacturing or purification of the product. 95.0 - 100.5 %
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR