Tetrahydrofuran 99,5 GC, corrected for water content Stabilised HPLC, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Butylene oxide, THF, Diethylene oxide

Stabilization: 75 - 400 ppm BHT

9440-03EA 76124.35 INR
9440-03 9440-50
Tetrahydrofuran 99,5 GC, corrected for water content Stabilised HPLC, J.T.Baker®
Tetrahydrofuran
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66
Melting Pt: –108.5 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: –21.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3 II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (by GC, corrected for water, exclusive of preservative) ≥99.5 %
Color (APHA) ≤10
Peroxide (as H₂O₂) ≤0.005 %
Stabilizer (BHT) 75 - 400 ppm
Water (by KF, coulometric) ≤0.010 %
Stabilized with approximately 0.01% Butylated Hydroxytoluene


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR