Acetonitrile, ultralow_water 99,5 GC, corrected for water content, BakerDRY™, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

9035-12EA 16224.32 INR
9035-12 9035-50 9035-98 9035-10 9035-K1 9035-K6
Acetonitrile, ultralow_water 99,5 GC, corrected for water content, BakerDRY™, J.T.Baker®
Acetonitrile

Specifically manufactured for use in organic, organometallic, and oligonucleotide synthesis requiring low water solvents.

Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6
Melting Pt: –45.7 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₃CN) (by GC, corrected for water) ≥99.5 %
Appearance Passes Test
ACS - Color (APHA) ≤10
ACS - Residue after Evaporation ≤1 ppm
ACS - Titrable Acid (µeq/g) ≤0.8
ACS - Titrable Base (µeq/g) ≤0.6
ACS - Water (by KF, coulometric) ≤10.0 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR