Potassium heptaiodobismuthate 6,1% aqueous L(+)-tartaric acid solution with potassium iodide Reag. Ph. Eur. 1070601

Supplier: VWR Chemicals

87913.270EA 0 INR
87913.270
Potassium heptaiodobismuthate 6,1% aqueous L(+)-tartaric acid solution with potassium iodide Reag. Ph. Eur. 1070601
Potassium heptaiodobismuthate

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: BiI₇K₄
MW: 1253.71 g/mol
CAS Number: 41944-01-8
EINECS: 255-595-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR