Phenol red : ammonium sulphate (0.002 : 0.005) % aqueous solution Reag. Ph. Eur. 1063603, buffered pH 4.7

Supplier: VWR Chemicals

87889.260EA 0 INR
87889.260
Phenol red : ammonium sulphate (0.002 : 0.005) % aqueous solution Reag. Ph. Eur. 1063603, buffered pH 4.7
Phenol red : ammonium sulphate

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: C₁₉H₁₃NaO₅S
Storage Temperature: Ambient
MDL Number: MFCD00066901
EINECS: 252-057-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR