Ninhydrin and stannous chloride reagent R1 (a+b) Reag. Ph. Eur. 1058302

Supplier: VWR Chemicals

87876.180EA 0 INR
87876.180
Ninhydrin and stannous chloride reagent R1 (a+b) Reag. Ph. Eur. 1058302
Ninhydrin and stannous chloride reagent

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Storage Temperature: Ambient
UN: 1188
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR