Ammonium heptamolybdate 10% aqueous solution Reag. Ph. Eur. 1005703

Supplier: VWR Chemicals

87771.290EA 0 INR
87771.290
Ammonium heptamolybdate 10% aqueous solution Reag. Ph. Eur. 1005703
Ammonium heptamolybdate solutions

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: (NH₄)₆Mo₇O₂₄·4H₂O
MW: 1235.86 g/mol
MDL Number: MFCD00167059
CAS Number: 12054-85-2
EINECS: 234-722-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR