VWR®, Manganese standard solution, 1000 mg/L Mn in dil. nitric acid AVS TITRINORM standard for AAS
Supplier: VWR Chemicals
86691.260EA 10241.19 INR
86691.260 86691.180
VWR®, Manganese standard solution, 1000 mg/L Mn in dil. nitric acid AVS TITRINORM standard for AAS
Standards Cationic Standards Manganese Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%