VWR® AVS TITRINORM, Lanthanum Standard Solution, 1000 mg/L La in dil. Nitric Acid AVS TITRINORM Standard for AAS
Supplier: VWR Chemicals
86687.180EA 6517.12 INR
86687.180 86687.260
VWR® AVS TITRINORM, Lanthanum Standard Solution, 1000 mg/L La in dil. Nitric Acid AVS TITRINORM Standard for AAS
Standards Cationic Standards Lanthanum Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%