VWR® AVS TITRINORM, Iron Standard Solution, 1000 mg/L Fe in 2% Hydrochloric Acid AVS TITRINORM Standard for AAS
Supplier: VWR Chemicals
86677.260EA 11014.12 INR
86677.260 86677.180
VWR® AVS TITRINORM, Iron Standard Solution, 1000 mg/L Fe in 2% Hydrochloric Acid AVS TITRINORM Standard for AAS
Standards Cationic Standards Iron Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%
Order Now
SPECIFICATIONS
Environmentally Preferable